HOLLOW CATHODE LAMP PWR SUP
Catalog #: CA11019-148
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
$260.09 to $2,400.95
Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
$224.36
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, HiPerSolv CHROMANORM®, super gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$741.97 to $47,885.70
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
$156.54 to $571.97