ADP KIT 37MM LAMPS-PE AA 9PIN
Catalog #: CA11019-136
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
146,85 $ to 576,51 $
Reinforced Plastic Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
433,11 $ to 433,11 $