SUPER LAMP 10V ARSENIC
Catalog #: CA11019-076
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
881,80 $ to 2 807,56 $
Sodium carbonate, anhydrous ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
129,84 $ to 1 081,96 $
Accessories for Advanced® Multi-Sample Osmometer, Model 2020, Advanced Instruments
Supplier: Advanced Instruments
1 223,38 $
Orion™ ROSS® Sure-Flow® Epoxy Combination pH Electrode, Thermo Scientific
Supplier: Thermo Fisher Scientific
2 940,02 $
Sol-Care® Safety Winged Blood Collection Needles, Sol M
Supplier: SOL-MILLENNIUM MEDICAL
84,04 $ to 651,48 $
Micromesh™ Biopsy Cassettes in Quickload™ Sleeves, Simport Scientific
Supplier: Simport Scientific
942,84 $