SUPER LAMP 3V CODED GBC NICKEL
Catalog #: CA11019-060
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$135.75 to $468.77
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
$1,313.27 to $5,149.78
Silver nitrate 0.1 N in aqueous solution volumetric solution, traceable to NIST Standard Reference Material (SRM)
Supplier: VWR
$347.81 to $1,296.17