SUPER LAMP 3V THALLIUM
Catalog #: CA11019-044
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
194,14 $ to 694,12 $
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
109,33 $ to 364,87 $
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
1 313,27 $ to 5 149,78 $