DEUTERIUM LAMP 10 V
Catalog #: CA11019-010
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
146,85 $ to 576,51 $
Iodine standardised solution 0.01 N in aqueous solution
Supplier: Thermo Scientific Chemicals
172,99 $
Sulfuric acid ≥95%, BAKER INSTRA-ANALYZED® for trace metal analysis low Selenium, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
483,15 $ to 1 605,80 $
Media/Lab Bottles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
103,73 $ to 117,47 $