DEUTERIUM LAMP 10 V
Catalog #: CA11019-006
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, HiPerSolv CHROMANORM®, super gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
741,97 $ to 47 885,70 $
Methanol ≥99.8%, GR ACS, meets analytical specification for testing USP/NF monographs, Supelco®
Supplier: MilliporeSigma
578,24 $ to 2 015,51 $
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
635,20 $ to 2 541,50 $
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15 × 100 mm plate, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
42,99 $