HCL 1.5 MULTI CO/CU/NI/ZN
Catalog #: CA11018-970
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
VWR® Prefilled Histology Containers, 10% Neutral Buffered Formalin (NBF)
Supplier: VWR International
$587.19
Hydrochloric acid 36.5 - 38.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$655.42 to $655.42
Sealing Films, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$192.55 to $218.23
Molecular sieve A4 (0.4 nm, 4 Å) ≥99%, pellets, Ø 3-4mm (0.12-0.16in)
Supplier: Thermo Scientific Chemicals
$75.71
Hydrion™ pH Test Paper Refills for Dispensers, Micro Essential Laboratory
Supplier: Micro Essential
$62.11 to $568.53