HCL 1.5 MULTI FE/ZN
Catalog #: CA11018-952
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Hydrochloric acid 36.5 - 38.0% ACS, NF, FCC, VWR Chemicals BDH®
Supplier: BDH
1 250,99 $ to 4 028,92 $
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
214,30 $ to 2 349,04 $
Tissue Marking Dyes (TMD™), TBS®, General Data
Supplier: General Data Healthcare
540,66 $ to 2 027,09 $
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
146,85 $ to 576,51 $
Sulfuric acid ≥95%, BAKER INSTRA-ANALYZED® for trace metal analysis low Selenium, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
483,15 $ to 1 605,80 $