HCL 1.5 MULTI AL/SI/TI
Catalog #: CA11018-940
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Hydrochloric acid 34 - 37%, OmniTrace® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
695,79 $
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
775,79 $ to 2 413,44 $
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
500,19 $ to 1 790,36 $
uTIP™ CleanPak™ Pipette Tip Reload System for Universal Fit Pipettes, Biotix Inc
Supplier: Biotix
81,72 $ to 326,77 $