HCL 1.5 MULTI CO/FE/NI/ZN
Catalog #: CA11018-906
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
Supplier: BD
$46.00 to $367.00
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
Supplier: Ricca Chemical
$205.73
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51