HCL 1.5 MULTI AL/CD/NI
Catalog #: CA11018-854
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Potassium nitrate ≥99.0%, crystallised ACS, VWR Chemicals BDH®
Supplier: VWR International
$179.52 to $927.60
Potassium chloride 99.0-100.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
$294.59 to $1,037.96
Conductivity Standards, Potassium Chloride (KCl) Solutions, 10 to 500 µS/cm, Thermo Scientific
Supplier: Thermo Scientific Chemicals
$71.01
Tryptic Soy Broth (TSB) with Lecithin and Tween®, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
$332.86
Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® ACS aldehyde-free, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$330.55 to $1,952.15