HCL 1.5 MULTI CR/FE/MN/NI
Catalog #: CA11018-840
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Water, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$277.09 to $740.95
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: Air-Tite
$105.11 to $2,771.71
Potassium nitrate ≥99.0%, crystallised ACS, VWR Chemicals BDH®
Supplier: VWR International
$179.52 to $927.60
Methanol ≥99.9%, B&J Brand™ LC-MS for LC-MS, for HPLC, Burdick & Jackson™
Supplier: Burdick & Jackson
$205.37 to $821.51
Sodium hydroxide 3 mol/l (3 N) in aqueous solution analytical reagent
Supplier: Thermo Scientific Chemicals
$251.20