HCL 1.5 MULTI CA/ZN
Catalog #: CA11018-828
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Tetrahydrofuran, anhydrous ≥99.8%, AcroSeal™ BHT-free, over molecular sieves
Supplier: Thermo Scientific Chemicals
$202.75
Hydrochloric acid 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$169.41 to $781.78
Nitric acid 67 - 70% (w/w), BAKER INSTRA-ANALYZED® Plus for trace metal analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$448.81
uTIP™ CleanPak™ Pipette Tip Reload System for Universal Fit Pipettes, Biotix Inc
Supplier: Biotix
$81.72 to $326.77