HCL 1.5 MULTI CU/FE/NI
Catalog #: CA11018-826
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Target™ All-Plastic Disposable Luer-Slip Syringes, Thermo Scientific
Supplier: Thermo Scientific
154,50 $ to 1 286,33 $
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
109,33 $ to 364,87 $
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
194,14 $ to 694,12 $
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
377,44 $ to 992,90 $