HCL 1.5 MULTI AG/CU/FE
Catalog #: CA11018-810
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Chloroform ≥99.8% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
558,88 $ to 2 230,71 $
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
146,85 $ to 576,51 $
Sodium hypochlorite 12.5% in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
99,85 $ to 385,36 $
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Supplier: Ricca Chemical
200,31 $ to 2 030,49 $
Sodium chloride 0.9% (w/v) in aqueous solution isotonic saline, suitable for use as blood bank saline
Supplier: Ricca Chemical
333,21 $
Brilliant II SYBR® MM with ROX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
615,00 $
Sodium chloride ≥99.0% (by argentometric titration), crystals, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
116,87 $ to 467,59 $