HCL 1.5 MULTI MO/SI
Catalog #: CA11018-800
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
$1,313.27 to $5,149.78
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$684.63 to $2,738.51
Listeria Palcam Selective Enrichment Broth Base Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
$663.68 to $3,804.95