HCL 1.5 MULTI CU/FE/MN/ZN
Catalog #: CA11018-764
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Benzyl alcohol ≥97%, AR®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$282.58 to $3,308.57
Sodium hydroxide 3 mol/l (3 N) in aqueous solution analytical reagent
Supplier: Thermo Scientific Chemicals
$251.20
ONETAQ® QUICK-LOAD® 2X Master Mix with Standard Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
$70.00