HCL 1.5 MULTI CR/FE/NI
Catalog #: CA11018-750
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$881.80 to $2,807.56
Sodium bisulfite ≥58.5%, granular, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$161.78 to $647.12
Exel International® Insulin Syringes with Fixed Needles, Air-Tite Products
Supplier: Air-Tite
$36.99 to $141.85