HCL 1.5 MULTI CR/FE/MN
Catalog #: CA11018-748
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
233,35 $ to 2 174,33 $
Masterflex® I/P® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
466,10 $
SystemSURE Plus™ Luminometer, ATP Hygiene Monitoring System, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
1 090,88 $
Sodium hypochlorite (11 - 15% Cl₂) available chlorine in aqueous solution
Supplier: Thermo Scientific Chemicals
136,01 $
Ammonium cerium(IV) nitrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
1 319,51 $ to 12 019,12 $
Color Standard, USP / EP Yellow Primary Solution, Ferric Chloride Solution, Ricca Chemical Company
Supplier: Ricca Chemical
508,14 $