HCL 1.5 MULTI CO/CR/CU/MN/NI
Catalog #: CA11018-700
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$599.10 to $2,396.15
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
$131.08 to $405.43
CAPINSERT™ for Cryovial® Tubes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$41.93 to $52.36