HCL 1.5 MULTI CD/PB
Catalog #: CA11018-692
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Potassium Standard, 1 ml = 0.1 mg K, 100 ppm K, Ricca Chemical Company
Supplier: Ricca Chemical
$118.85
Choline chloride 98.0-100.5% (by anhydrous basis), BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$149.28
Molecular sieve A4 (0.4 nm, 4 Å), BAKER ANALYZED®, activated, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$247.90
Hydrofluoric acid 47 - 51% (w/w), BAKER INSTRA-ANALYZED® Plus for trace metal analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$311.43
Orthophosphoric acid ≥85.0% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$285.47 to $1,142.06