HCL 1.5 MULTI AL/SI/TI
Catalog #: CA11018-686
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
214,30 $ to 2 349,04 $
VRB (Violet Red Bile) Agar, Fluorocult®, Dehydrated Culture Media, MilliporeSigma
Supplier: MilliporeSigma
1 692,83 $
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
78,90 $ to 874,91 $
Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
Supplier: BD
46,00 $ to 367,00 $
Chloroform ≥99.8% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
558,88 $ to 2 230,71 $
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
99,02 $ to 485,35 $