HCL 1.5 MULTI CR/FE/MN/TI
Catalog #: CA11018-670
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Ethylene glycol ≥99%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
321,66 $ to 979,57 $
Reagent alcohol absolute 94.0-96.0%, AR® ACS (denatured with 5% isopropanol and 5% methanol), Macron Fine Chemicals™
Supplier: VWR International
113,32 $ to 1 359,75 $
Molecular sieve A4 (0.4 nm, 4 Å) ≥99%, pellets, Ø 3-4mm (0.12-0.16in)
Supplier: Thermo Scientific Chemicals
75,71 $
Orthophosphoric acid ≥85.0% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
285,47 $ to 1 142,06 $