HCL 1.5 MULTI AG/CD/PB/ZN
Catalog #: CA11018-652
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Hydrochloric acid 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
169,41 $ to 781,78 $
Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® ACS aldehyde-free, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
330,55 $ to 1 952,15 $
Conductivity Standards, Potassium Chloride (KCl) Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
222,01 $
Blunt Needle Luer Stub Adaptors, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
69,11 $
Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor, Thermo Scientific
Supplier: Thermo Fisher Scientific
760,08 $
Sodium bisulfite ≥90% FCC, purified, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
105,59 $