HCL 1.5 MULTI CU/MO/ZN
Catalog #: CA11018-628
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Hydrochloric acid 36.5 - 38.0%, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$103.36 to $1,240.08
Sodium acetate trihydrate 99.0-101.0% (dried basis), crystals USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$1,159.74 to $3,133.57
EcoRI, Restriction Enzymes, from <i>E.coli</i>, New England Biolabs
Supplier: New England Biolabs (NEB)
$100.00
Carbon Steel Surgical Blades, Non Sterile, Cincinnati Surigcal
Supplier: CINCINNATI SURGICAL CO.
$131.23
Copper ≥99.9% (metals basis), shot 1-10 mm (0.04-0.4 in)
Supplier: Thermo Scientific Chemicals
$106.00