HCL 1.5 MULTI CD/ZN
Catalog #: CA11018-608
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Tissue Marking Dyes (TMD™), TBS®, General Data
Supplier: General Data Healthcare
540,66 $ to 2 027,09 $
Ethanol denatured (denatured with 5% isopropanol and 5% methanol), VWR Chemicals BDH®
Supplier: VWR International
147,27 $ to 464,44 $
Cerium(IV) sulfate solution 0.1 N, VWR Chemicals BDH®
Supplier: VWR International
470,52 $ to 5 298,64 $
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
346,72 $