HCL 1.5 MULTI CA/MG/SI
Catalog #: CA11018-606
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
881,80 $ to 2 807,56 $
Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
415,40 $
Micrewtube® Microcentrifuge Tube with Screw Cap, Simport Scientific
Supplier: Simport Scientific
226,19 $
Dichloromethane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
285,68 $ to 3 428,09 $