HCL 1.5 MULTI AG/PB
Catalog #: CA11018-594
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
635,20 $ to 2 541,50 $
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
96,73 $ to 485,35 $
HistoTainer™ Prefilled Specimen Containers, with 10% Neutral Buffered Formalin, Simport Scientific
Supplier: Simport Scientific
131,44 $
Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
Supplier: BD
46,00 $ to 367,00 $