HCL 1.5 MULTI CR/SE
Catalog #: CA11018-588
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Masterflex® L/S® Precision Pump Tubing, FDA-Compliant Viton®, Avantor®
Supplier: VWR International
268,60 $
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
775,79 $ to 2 413,44 $
Chloroform ≥99.8% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
558,88 $ to 2 230,71 $
Potassium bromate ≥99.8% (by iodometry), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
291,16 $
n-Heptane ≥99%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
359,51 $ to 1 103,97 $
Tryptic Soy Agar (TSA), Soybean Casein Digest Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
39,00 $