HCL 1.5 MULTI AU/CU
Catalog #: CA11018-496
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Sodium hydroxide 50% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
684,53 $
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
233,35 $ to 2 174,33 $
Hydrofluoric acid 47 - 51% (w/w), BAKER INSTRA-ANALYZED® Plus for trace metal analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
311,43 $