PERKIN ELM 4PIN LUMNA THALLIUM
Catalog #: CA11018-454
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
635,20 $ to 2 541,50 $
Acetonitrile ≥99.9% (by GC) for HPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
885,46 $ to 3 541,82 $
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
1 313,27 $ to 5 149,78 $
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
881,80 $ to 2 807,56 $