PERKIN ELM 4PIN LUMINA NIOBIUM
Catalog #: CA11018-416
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Hexane (mixture of isomers) ≥98.5% ACS for HPLC, Supelco®
Supplier: MilliporeSigma
649,44 $ to 2 496,86 $
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
109,33 $ to 364,87 $
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
1 313,27 $ to 5 149,78 $