PERKIN ELM 4PIN LUMINA BARIUM
Catalog #: CA11018-354
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$194.14 to $694.12
Sodium hypochlorite 12.5% in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$99.85 to $385.36
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$500.19 to $1,790.36
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
$99.02 to $485.35