PERKIN ELMER 9PIN TERBIUM
Catalog #: CA11018-300
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Single-Use Syringe/BD PrecisionGlide™ Needle Combinations, Sterile, BD Medical
Supplier: BD
46,00 $ to 367,00 $
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
635,20 $ to 2 541,50 $
Hydrion® Single Roll Dispenser, pH 1.0 to 12.0, Micro Essential Laboratory®
Supplier: Micro Essential
137,62 $
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
775,79 $ to 2 413,44 $
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
500,19 $ to 1 790,36 $
Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
667,83 $