PERKIN ELMER 9PIN RUBIDIUM
Catalog #: CA11018-278
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
154,66 $
Potassium dihydrogen phosphate ≥99.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
1 033,55 $ to 4 052,62 $
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15 × 100 mm plate, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
42,99 $
Retriever Thermal Slide Processr, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
462,20 $ to 483,01 $