PERKIN ELMER 9PIN LANTHANUM
Catalog #: CA11018-242
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
VACUETTE® Blood Collection Tubes, without Additive, Greiner bio-one
Supplier: Greiner Bio-One
$13.18 to $316.03
Zinc sulfate heptahydrate ≥99%, granular USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$273.06
Conductivity Standards, Potassium Chloride (KCl) Solutions, Thermo Scientific
Supplier: Thermo Scientific Chemicals
$78.00
Pierce™ DNA Extraction Buffer, Phenol:Chloroform and Tris Buffer, Thermo Scientific
Supplier: Thermo Scientific
$628.97
Dichloromethane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$471.36 to $2,828.23
Orthophosphoric acid ≥85.0% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$285.47 to $1,142.06