PERKIN ELMER 9PIN ERBIUM
Catalog #: CA11018-220
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Target™ All-Plastic Disposable Luer-Slip Syringes, Thermo Scientific
Supplier: Thermo Scientific
$89.28 to $679.72
Barium chloride dihydrate ≥99.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
$184.95 to $1,807.94
Sodium hydroxide 3 mol/l (3 N) in aqueous solution analytical reagent
Supplier: Thermo Scientific Chemicals
$251.20
Glycerine ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$1,139.76 to $4,558.98
Flow cells for MinION devices, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
$7,900.00