PERKIN ELMER 9PIN BERYLLIUM
Catalog #: CA11018-202
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
170,25 $ to 676,42 $
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
131,08 $ to 405,43 $
Hydrochloric acid 34 - 37%, OmniTrace® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
309,76 $ to 2 237,83 $
Borate Buffer, pH 9.5, for Ammonia and Organic Nitrogen Analysis, Ricca Chemical Company
Supplier: Ricca Chemical
162,71 $