HCL THRM FSHR CODED SCANDIUM
Catalog #: CA11018-144
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
109,33 $ to 364,87 $
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.9%, HiPerSolv CHROMANORM®, super gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
741,97 $ to 47 885,70 $
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
377,44 $ to 992,90 $
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
194,14 $ to 694,12 $