HCL THRM FSHR CODED MANGANESE
Catalog #: CA11018-112
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Formaldehyde 37% in aqueous solution stabilized, VWR®, Reagent Grade
Supplier: VWR International
77,47 $ to 576,55 $
Hydrochloric acid 50% (v/v) (1+1), VWR Chemicals BDH®
Supplier: VWR International
200,77 $ to 799,37 $
Fluoride Standards, 1 ml = 1 mg F-, 1000 ppm F-, RICCA Chemical Company
Supplier: Ricca Chemical
166,86 $
Feather® Safeshield™ Scalpels, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76,85 $ to 87,03 $