HCL THRM FSHR CODED ERBIUM
Catalog #: CA11018-080
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
775,79 $ to 2 413,44 $
Dichloromethane ≥99.8% stabilized, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
389,38 $ to 1 030,72 $
Potassium, Single Element ICP and ICP/MS Certified Reference Standards, Enhanced Packaging, ARISTAR®, , VWR® BDH®
Supplier: VWR International
338,38 $
Insulin Syringes with Permanently Attached Needles, BD Medical
Supplier: Cardinal Health
99,02 $ to 485,35 $