HCL 1.5 CODED THORIUM
Catalog #: CA11018-018
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Hydrogen peroxide 30% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
214,30 $ to 2 349,04 $
Aquastar™ Coulometric and Volumetric Reagents and Solvents, MilliporeSigma
Supplier: MilliporeSigma
386,44 $ to 1 545,78 $
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
377,44 $ to 992,90 $
Nitric acid 67 - 70%, OmniTrace® for trace metal analysis, Supelco®
Supplier: MilliporeSigma
1 209,55 $ to 4 743,09 $