HCL 1.5 CODED THALLIUM
Catalog #: CA11018-016
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Ethanol denatured ≥80% (denatured with methanol and ethyl acetate)
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$599.10 to $2,396.15
Alcohol ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
$131.08 to $405.43
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15 × 100 mm plate, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
$42.99