HCL 1.5 CODED RHODIUM
Catalog #: CA11017-954
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Glycerine ≥99.7%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
881,80 $ to 2 807,56 $
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
BD Vacutainer® SPC Plus Blood Collection Tubes with BD Hemogard™ Closures, BD Medical
Supplier: BD
124,23 $ to 1 196,70 $
Flow cells for MinION devices, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
7 900,00 $
Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Supplier: MilliporeSigma
178,29 $