HCL 1.5 CODED RHENIUM
Catalog #: CA11017-952
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Ethylene glycol ≥99.0%, Semi Grade for the electronics industry, VWR Chemicals BDH®
Supplier: BDH
$122.61 to $485.99
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Supplier: Ricca Chemical
$200.31 to $2,030.49
Hydrochloric acid 32 - 35%, OmniTrace Ultra™ for trace metal analysis, Supelco®
Supplier: MilliporeSigma
$1,264.56
Sodium hydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$231.20 to $2,774.16
Pierce SnakeSkin™ Pleated Dialysis Tubing, MWCO 3500 to 10000, Thermo Scientific
Supplier: Thermo Scientific
$439.41
Flow cells for MinION devices, Oxford Nanopore Technologies
Supplier: Oxford Nanopore Technologies
$7,900.00