HCL 1.5 CODED NICKEL
Catalog #: CA11017-938
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Hydrochloric acid 50% (v/v) (1+1), VWR Chemicals BDH®
Supplier: VWR International
99,45 $ to 1 061,29 $
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
64,40 $ to 734,93 $
Hydrochloric acid 36.5 - 38.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
655,42 $ to 655,42 $
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics
316,00 $ to 5 024,40 $