HCL 1.5 CODED MOLYBDENUM
Catalog #: CA11017-934
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Supplier: Air-Tite
110,86 $ to 918,90 $
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
377,44 $ to 992,90 $
Reagent alcohol ACS (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
260,48 $ to 788,04 $
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
684,63 $ to 2 738,51 $
Sodium phosphate dibasic, anhydrous ≥99.0%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: MilliporeSigma
622,34 $
Scalpel Handles; Feather®, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
54,46 $