HCL 1.5 CODED MANGANESE
Catalog #: CA11017-930
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Sodium hydroxide 40% (w/w) in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
146,85 $ to 576,51 $
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
775,79 $ to 2 413,44 $
VWR® PurFlock Ultra 6" Sterile Standard Flock Swab with Polystyrene Handle, 80 mm Breakpoint
Supplier: VWR International
31,15 $ to 302,35 $
Minimate™ EVO Tangential Flow Filtration System and Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
7 496,08 $
Thermo-Flask Stainless Steel, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
2 203,85 $