HCL 1.5 HOLMIUM
Catalog #: CA11017-812
Supplier: PHOTRON CA
Customers Who Bought This Also Bought
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Supplier: VWR International
$109.33 to $364.87
Diethyl ether ≥98% stabilized ACS, VWR Chemicals BDH®
Supplier: VWR International
$775.79 to $2,413.44
Methanol ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC, VWR Chemicals BDH®
Supplier: VWR International
$377.44 to $992.90
Sodium hypochlorite 12.5% in aqueous solution, VWR Chemicals BDH®
Supplier: VWR International
$99.85 to $385.36
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$500.19 to $1,790.36